Gene Transcripts

Gene Symbol
UBA6
Entrez Gene ID
55236
Gene Name
ubiquitin like modifier activating enzyme 6
Gene Aliases
E1-L2, MOP-4, UBE1L2
Chromosome Location
Chr. 4: 67615761 - 67701171 on Build GRCh38
UniGene ID
Hs.212774
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018227.5 NP_060697.4 13, 14 1153
XM_017008359.1 XP_016863848.1 13, 14 1130
GenBank mRNA AB014773.1 - 13, 14 1101
AK094164.1 - 5, 6 359
AK314371.1 - 13, 14 1130
AL832015.2 - 13, 14 1122
AL832458.2 - 13, 14 1122
BC126484.1 - 13, 14 1148 8992200
BC126486.1 - 13, 14 1148 8992202
EF623993.1 - 13, 14 1101

View Full Product Details