Gene Transcripts

Gene Symbol
TTC29
Entrez Gene ID
83894
Gene Name
tetratricopeptide repeat domain 29
Gene Aliases
NYD-SP14, TBPP2A
Chromosome Location
Chr. 4: 146706638 - 146945886 on Build GRCh38
UniGene ID
Hs.378893
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300761.2 NP_001287690.1 7 818
NM_001317806.1 NP_001304735.1 6 717
NM_031956.3 NP_114162.2 6 717
NR_133922.1 6 717
XM_005263270.1 XP_005263327.1 7 800
XM_006714335.1 XP_006714398.1 7 800
XM_006714336.1 XP_006714399.1 7 800
XM_011532310.2 XP_011530612.1 7 800
GenBank mRNA AF345910.1 - 6 668
AK093145.1 - 7 764
AK225725.1 - 6 664
AK301807.1 - 6 643
AK314066.1 - 6 713
BC024193.2 - 6 659 4825866
EF432564.1 - 4 486

View Full Product Details