Gene Transcripts

Gene Symbol
RRP36
Entrez Gene ID
88745
Gene Name
ribosomal RNA processing 36
Gene Aliases
C6orf153, dJ20C7.4
Chromosome Location
Chr. 6: 43021647 - 43029599 on Build GRCh38
UniGene ID
Hs.309231
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033112.2 NP_149103.1 3 336
GenBank mRNA AF161371.1 - 2 92
AK313816.1 - 3 346
BC006293.2 - 3 330
BC011933.2 - 3 336 4300313

View Full Product Details