Gene Transcripts
- Gene Symbol
- SLC44A3
- Entrez Gene ID
- 126969
- Gene Name
- solute carrier family 44 member 3
- Gene Aliases
- CTL3
- Chromosome Location
- Chr. 1: 94820342 - 94895247 on Build GRCh38
- UniGene ID
- Hs.483423
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001114106.2 |
NP_001107578.1
|
7, 8 |
870 |
|
NM_001258340.1 |
NP_001245269.1
|
7, 8 |
870 |
|
NM_001258341.1 |
NP_001245270.1
|
7, 8 |
774 |
|
NM_001258342.1 |
NP_001245271.1
|
6, 7 |
762 |
|
NM_001258343.1 |
NP_001245272.1
|
6, 7 |
666 |
|
NM_001301079.1 |
NP_001288008.1
|
6, 7 |
709 |
|
NM_152369.4 |
NP_689582.2
|
6, 7 |
805 |
|
XM_005270440.2 |
XP_005270497.1
|
7, 8 |
765 |
|
XM_005270441.2 |
XP_005270498.1
|
7, 8 |
807 |
|
XM_005270442.1 |
XP_005270499.1
|
7, 8 |
861 |
|
XM_011540632.2 |
XP_011538934.1
|
6, 7 |
734 |
|
XM_011540633.2 |
XP_011538935.1
|
6, 7 |
572 |
|
XM_011540635.2 |
XP_011538937.1
|
7, 8 |
861 |
|
XM_011540636.2 |
XP_011538938.1
|
7, 8 |
765 |
|
XM_017000262.1 |
XP_016855751.1
|
6, 7 |
747 |
|
XM_017000263.1 |
XP_016855752.1
|
7, 8 |
807 |
|
XM_017000264.1 |
XP_016855753.1
|
7, 8 |
861 |
|
XM_017000265.1 |
XP_016855754.1
|
7, 8 |
765 |
|
XM_017000266.1 |
XP_016855755.1
|
7, 8 |
861 |
|
GenBank mRNA |
AK300193.1 |
- |
6, 7 |
603 |
|
AK301288.1 |
- |
6, 7 |
611 |
|
AK302381.1 |
- |
4, 5 |
454 |
|
AK303906.1 |
- |
6, 7 |
741 |
|
AK313574.1 |
- |
7, 8 |
758 |
|
AK316123.1 |
- |
8, 9 |
715 |
|
AL530322.3 |
- |
7, 8 |
862 |
|
AL540829.3 |
- |
6, 7 |
762 |
|
AY358659.1 |
- |
3, 4 |
324 |
|
BC033858.1 |
- |
7, 8 |
833 |
|
BC053877.1 |
- |
6, 7 |
728 |
5760394 |
BX648225.1 |
- |
6, 7 |
734 |
|
View Full Product Details