Gene Transcripts
- Gene Symbol
- TEPSIN
- Entrez Gene ID
- 146705
- Gene Name
- TEPSIN, adaptor related protein complex 4 accessory protein
- Gene Aliases
- C17orf56, ENTHD2
- Chromosome Location
- Chr. 17: 81228277 - 81239092 on Build GRCh38
- UniGene ID
- Hs.631761
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_144679.2 |
NP_653280.1
|
3, 4 |
268 |
|
XM_005257066.2 |
XP_005257123.1
|
4, 5 |
862 |
|
XM_005257067.2 |
XP_005257124.1
|
3, 4 |
268 |
|
XM_006721709.3 |
XP_006721772.1
|
3, 4 |
275 |
|
XM_006721712.2 |
XP_006721775.1
|
3, 4 |
706 |
|
XM_011524355.1 |
XP_011522657.1
|
4, 5 |
868 |
|
XM_011524356.1 |
XP_011522658.1
|
4, 5 |
868 |
|
XM_011524357.1 |
XP_011522659.1
|
3, 4 |
896 |
|
XM_011524358.2 |
XP_011522660.1
|
4, 5 |
797 |
|
XM_017024202.1 |
XP_016879691.1
|
4, 5 |
1109 |
|
XM_017024203.1 |
XP_016879692.1
|
5, 6 |
756 |
|
XM_017024204.1 |
XP_016879693.1
|
4, 5 |
1309 |
|
GenBank mRNA |
AK056090.1 |
- |
3, 4 |
232 |
|
AK127221.1 |
- |
1, 2 |
1421 |
|
AK128728.1 |
- |
4, 5 |
1581 |
|
BC064483.1 |
- |
3, 4 |
229 |
5430837 |
DB122720.1 |
- |
3, 4 |
268 |
|
View Full Product Details