Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001100600.1 | NP_001094070.1 | 1 | 220 | |
NM_001270375.1 | NP_001257304.1 | 1 | 220 | ||
NM_198403.3 | NP_940685.3 | 1 | 220 | ||
NR_072989.1 | 1 | 220 | |||
GenBank mRNA | AK094524.1 | - | 1 | 219 | |
BC037881.1 | - | 1 | 171 | 5274201 | |
BC067905.1 | - | 1 | 220 | 5312325 |