Gene Transcripts

Gene Symbol
GYPC
Entrez Gene ID
2995
Gene Name
glycophorin C (Gerbich blood group)
Gene Aliases
CD236, CD236R, GE, GPC, GPD, GYPD, PAS-2, PAS-2'
Chromosome Location
Chr. 2: 126655935 - 126696675 on Build GRCh38
UniGene ID
Hs.59138
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256584.1 NP_001243513.1 5 1381
NM_002101.4 NP_002092.1 4 563
NM_016815.3 NP_058131.1 3 506
XM_006712460.3 XP_006712523.1 4 431
GenBank mRNA AK312032.1 - 4 351
BC104246.1 - 4 364 40033899
BC104247.1 - 4 364 40033903
BC106051.1 - 4 395 4707035
CB992018.1 - 3 291
M11802.1 - 4 234
M28335.1 - 4 228
M36284.1 - 4 234
X12496.1 - 4 354
X51973.1 - 3 177

View Full Product Details