Gene Transcripts

Gene Symbol
MAGOH
Entrez Gene ID
4116
Gene Name
mago homolog, exon junction complex core component
Gene Aliases
MAGOH1, MAGOHA
Chromosome Location
Chr. 1: 53226892 - 53238610 on Build GRCh38
UniGene ID
Hs.421576
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002370.3 NP_002361.1 1 210
GenBank mRNA AF035940.1 - 1 113
AF067173.1 - 1 136
AK297895.1 - 1 127
AK312113.1 - 1 146
BC018211.1 - 1 122 3861094
BX283982.1 - 1 210

View Full Product Details