Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012394.3 | NP_036526.2 | 3 | 275 | |
XM_011509624.2 | XP_011507926.1 | 4 | 288 | ||
GenBank mRNA | AF117237.1 | - | 3 | 231 | |
AF151065.1 | - | 3 | 215 | ||
AF165883.1 | - | 3 | 248 | ||
AK024531.1 | - | 3 | 255 | ||
AK315736.1 | - | 3 | 249 | ||
BC012464.1 | - | 3 | 244 | 4538003 | |
BC047042.1 | - | 3 | 241 | 5541226 | |
BI597083.1 | - | 3 | 308 |