Gene Transcripts

Gene Symbol
PSG7
Entrez Gene ID
5676
Gene Name
pregnancy specific beta-1-glycoprotein 7 (gene/pseudogene)
Gene Aliases
PSBG-7, PSG1, PSGGA
Chromosome Location
Chr. 19: 42924132 - 42937178 on Build GRCh38
UniGene ID
Hs.709203
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206650.1 NP_001193579.1 5 1127
NM_001290042.1 NP_001276971.1 6 1493
NM_002783.2 NP_002774.2 6 1493
GenBank mRNA BC030979.1 - 6 1488
BC136401.1 - 5 1047
U18467.1 - 6 1443

View Full Product Details