Gene Transcripts

Gene Symbol
SPRR2A
Entrez Gene ID
6700
Gene Name
small proline rich protein 2A
Gene Aliases
Chromosome Location
Chr. 1: 153056120 - 153057512 on Build GRCh38
UniGene ID
Hs.355542
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005988.2 NP_005979.1 2 251
GenBank mRNA AK311939.1 - 2 252
BC096108.1 - 1 224 40002232
BC096109.1 - 1 224 40002234
BC096110.1 - 1 224 40002235
BC096111.1 - 1 224 40002236
BC128049.1 - 2 234 40115066

View Full Product Details