Gene Transcripts

Gene Symbol
TOR2A
Entrez Gene ID
27433
Gene Name
torsin family 2 member A
Gene Aliases
TORP1
Chromosome Location
Chr. 9: 127731524 - 127735387 on Build GRCh38
UniGene ID
Hs.444106
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085347.2 NP_001078816.1 2, 3 484
NM_001134430.2 NP_001127902.1 2, 3 484
NM_130459.3 NP_569726.2 2, 3 484
GenBank mRNA AK027677.1 - 2, 3 419
AK075520.1 - 2, 3 427
AY358180.1 - 2, 3 451
CN263138.1 - 2, 3 486
DA430836.1 - 2, 3 460

View Full Product Details