Gene Transcripts
- Gene Symbol
- TTTY14
- Entrez Gene ID
- 83869
- Gene Name
- testis-specific transcript, Y-linked 14 (non-protein coding)
- Gene Aliases
- CYorf14, NCRNA00137, NCRNA00185, PRO2834, TTY14
- Chromosome Location
- Chr. Y: 18872501 - 19077547 on Build GRCh38
- UniGene ID
- Hs.138453
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF119903.1 |
- |
LIT2233 CYorf14 |
1 |
606 |
|
View Full Product Details