Gene Transcripts

Gene Symbol
SIRPG
Entrez Gene ID
55423
Gene Name
signal regulatory protein gamma
Gene Aliases
CD172g, SIRP-B2, SIRPB2, SIRPgamma, bA77C3.1
Chromosome Location
Chr. 20: 1629152 - 1686516 on Build GRCh38
UniGene ID
Hs.590883
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039508.1 NP_001034597.1 2 228
NM_018556.3 NP_061026.2 2 228
NM_080816.2 NP_543006.2 2 228
XM_005260749.3 XP_005260806.1 2 264
XM_011529286.2 XP_011527588.1 2 275
XM_011529287.2 XP_011527589.1 2 231
GenBank mRNA AB042624.1 - 2 244
AK292745.1 - 2 228
AY748247.1 - 2 163
AY748248.1 - 2 163
BC020629.1 - 2 205
BC064532.1 - 2 189 5588038
DA008008.1 - 2 231

View Full Product Details