Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020798.2 | NP_065849.1 | 6 | 1526 | |
XM_011545187.2 | XP_011543489.1 | 6 | 669 | ||
XM_011545188.2 | XP_011543490.1 | 4 | 452 | ||
XM_017018052.1 | XP_016873541.1 | 6 | 1523 | ||
XM_017018053.1 | XP_016873542.1 | 5 | 518 | ||
GenBank mRNA | AB037793.1 | - | 3 | 989 | |
AJ586137.1 | - | 5 | 1167 | ||
AK307875.1 | - | 5 | 752 | ||
BC131489.1 | - | 4 | 489 | 40038029 |