Gene Transcripts
- Gene Symbol
- TAS2R19
- Entrez Gene ID
- 259294
- Gene Name
- taste 2 receptor member 19
- Gene Aliases
- MSTP058, T2R19, T2R23, T2R48, TAS2R23, TAS2R48
- Chromosome Location
- Chr. 12: 11021619 - 11022620 on Build GRCh38
- UniGene ID
- Hs.687025
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF116728.1 |
- |
1 |
1329 |
|
View Full Product Details