Gene Transcripts

Gene Symbol
TIMM17A
Entrez Gene ID
10440
Gene Name
translocase of inner mitochondrial membrane 17 homolog A (yeast)
Gene Aliases
TIM17, TIM17A
Chromosome Location
Chr. 1: 201955491 - 201970661 on Build GRCh38
UniGene ID
Hs.20716
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006335.2 NP_006326.1 5 396
GenBank mRNA AF106622.1 - 5 367
AK023063.1 - 2 1570
AK315602.1 - 5 384
AU123184.1 - 5 396
BC000294.2 - 5 366 2819455
BC004439.1 - 5 366 2819455
BC007106.1 - 5 377 4284092
BC009784.1 - 5 388 3683125
BC013823.1 - 5 355
BC015098.1 - 5 394 3686369
BC016817.1 - 3 251
BC020833.1 - 5 374 4773149
X97544.1 - 5 373

View Full Product Details