Gene Transcripts
- Gene Symbol
- TEPSIN
- Entrez Gene ID
- 146705
- Gene Name
- TEPSIN, adaptor related protein complex 4 accessory protein
- Gene Aliases
- C17orf56, ENTHD2
- Chromosome Location
- Chr. 17: 81228277 - 81239092 on Build GRCh38
- UniGene ID
- Hs.631761
- Species
- Homo sapiens
- Gene Symbol
- LOC105371925
- Entrez Gene ID
- 105371925
- Gene Name
- uncharacterized LOC105371925
- Gene Aliases
- Chromosome Location
- Chr. 17: 81228707 - 81232209 on Build GRCh38
- UniGene ID
- Hs.620720
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL832593.1 |
- |
3 |
2523 |
|
View Full Product Details