Gene Transcripts

Gene Symbol
ZCCHC11
Entrez Gene ID
23318
Gene Name
zinc finger CCHC-type containing 11
Gene Aliases
PAPD3, TUT4
Chromosome Location
Chr. 1: 52423275 - 52553463 on Build GRCh38
UniGene ID
Hs.655407
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009881.2 NP_001009881.1 28 4571
NM_015269.2 NP_056084.1 28 4568
XM_005270676.2 XP_005270733.1 28 4580
XM_005270678.2 XP_005270735.1 28 4577
XM_005270679.2 XP_005270736.1 28 4565
XM_005270680.2 XP_005270737.1 28 4565
XM_005270681.2 XP_005270738.1 27 4466
XM_005270682.2 XP_005270739.1 26 4247
XM_005270683.2 XP_005270740.1 24 4214
XM_006710498.2 XP_006710561.1 28 4517
XM_006710499.3 XP_006710562.1 28 4524
XM_006710500.1 XP_006710563.1 27 4367
XM_011541097.1 XP_011539399.1 29 4602
XM_011541098.1 XP_011539400.1 28 4499
XM_011541099.1 XP_011539401.1 27 4286
XM_017000803.1 XP_016856292.1 28 4484
XM_017000804.1 XP_016856293.1 28 4484
XM_017000805.1 XP_016856294.1 27 4451
XM_017000806.1 XP_016856295.1 27 4352
XM_017000807.1 XP_016856296.1 27 4352
XM_017000808.1 XP_016856297.1 26 4232
XM_017000809.1 XP_016856298.1 26 4247
XM_017000810.1 XP_016856299.1 26 4232
XM_017000811.1 XP_016856300.1 24 4214
XM_017000812.1 XP_016856301.1 24 4211
XM_017000813.1 XP_016856302.1 24 4199
XM_017000814.1 XP_016856303.1 22 3881
GenBank mRNA AK025698.1 - 12 1225
BC131734.1 - 28 4463 40125005
BX439159.2 - 8 829
BX648248.1 - 1 3265
D83776.1 - 27 3933

View Full Product Details