Gene Transcripts

Gene Symbol
TMEM97
Entrez Gene ID
27346
Gene Name
transmembrane protein 97
Gene Aliases
MAC30
Chromosome Location
Chr. 17: 28319041 - 28328685 on Build GRCh38
UniGene ID
Hs.199695
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014573.2 NP_055388.2 1, 2 274
GenBank mRNA AK293331.1 - 1, 2 129
AK299511.1 - 1, 2 167
AK307678.1 - 1, 2 166
BC017362.2 - 1, 2 145
BC091504.1 - 1, 2 145 30379019
DA410183.1 - 1, 2 274
L19183.1 - 1, 2 168

View Full Product Details