Gene Transcripts

Gene Symbol
RADIL
Entrez Gene ID
55698
Gene Name
Rap associating with DIL domain
Gene Aliases
RASIP2
Chromosome Location
Chr. 7: 4799109 - 4883704 on Build GRCh38
UniGene ID
Hs.667336
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018059.4 NP_060529.4 10, 11 2473
GenBank mRNA AB058752.1 - 10, 11 2417
AK000997.1 - 6, 7 911
AK001186.1 - 7, 8 1456
AK294745.1 - 9, 10 2272
AK296325.1 - 9, 10 1994
AK302549.1 - 2465
AL136731.1 - 5, 6 704
BC004919.2 - 1 324
BC117317.1 - 9, 10 2359 40125868
BC126311.1 - 9, 10 2359 8992027
BC143526.1 - 9, 10 2359 9052035
EF560723.1 - 9, 10 2382

View Full Product Details