Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001033219.3 | NP_001028391.2 | 3 | 1116 | |
NM_001168255.1 | NP_001161727.1 | 2 | 194 | ||
GenBank mRNA | AK133269.1 | - | 3 | 1117 | |
AK170738.1 | - | 2 | 199 | ||
BC056501.1 | - | 3 | 713 | 5720940 | |
BC058722.1 | - | 2 | 88 | ||
BY200093.1 | - | 2 | 199 |