Gene Transcripts
- Gene Symbol
- Mrvi1
- Entrez Gene ID
- 17540
- Gene Name
- MRV integration site 1
- Gene Aliases
- BB115629, Irag, Ris1
- Chromosome Location
- Chr. 7: 110868257 - 110982908 on Build GRCm38
- UniGene ID
- Mm.250023
- Species
- Mus musculus
- Gene Symbol
- Gm16336
- Entrez Gene ID
- 100504184
- Gene Name
- predicted gene 16336
- Gene Aliases
- OTTMUSG00000033852
- Chromosome Location
- Chr. 7: 110904985 - 110905851 on Build GRCm38
- UniGene ID
- Mm.441327
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BE633821.1 |
- |
2 |
134 |
|
View Full Product Details