Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172517.2 | NP_766105.2 | 12 | 1486 | |
GenBank mRNA | AK049247.1 | - | 12 | 1473 | |
AK154782.1 | - | 12 | 1487 | ||
AK170520.1 | - | 12 | 1484 | ||
AK171177.1 | - | 12 | 1491 | ||
BC030892.1 | - | 6 | 687 | ||
BC057632.1 | - | 13 | 1359 | 4918318 | |
BC060186.1 | - | 1 | 43 |