Gene Transcripts

Gene Symbol
Rrp36
Entrez Gene ID
224823
Gene Name
ribosomal RNA processing 36 homolog (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 17: 46667442 - 46674451 on Build GRCm38
UniGene ID
Mm.45101
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144857.1 NP_659106.1 2 131
GenBank mRNA AK148164.1 - 164
AK148237.1 - 2 228
BC011248.1 - 2 131 4192711

View Full Product Details