Gene Transcripts

Gene Symbol
Trpa1
Entrez Gene ID
277328
Gene Name
transient receptor potential cation channel, subfamily A, member 1
Gene Aliases
Anktm1, TRPA1b
Chromosome Location
Chr. 1: 14872648 - 14919014 on Build GRCm38
UniGene ID
Mm.186329
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177781.4 NP_808449.1 11, 12 1397
GenBank mRNA AK045771.1 - 11, 12 1398
AY231177.1 - 11, 12 1370
BC120563.1 - 11, 12 1388 40129486
BC131963.1 - 11, 12 1388 40130240
KF460461.1 - 11, 12 1370

View Full Product Details