Gene Transcripts

Gene Symbol
Prss30
Entrez Gene ID
30943
Gene Name
protease, serine 30
Gene Aliases
2010015P21Rik, Disp, Disp1, Tmprss8
Chromosome Location
Chr. 17: 23972127 - 23980572 on Build GRCm38
UniGene ID
Mm.142568
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013921.3 NP_038949.2 3 205
GenBank mRNA AJ243866.1 - 3 168
BC010970.1 - 3 198 4216644
BC040348.1 - 3 205 4206523

View Full Product Details