Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025706.3 | NP_079982.3 | 13 | 1532 | |
GenBank mRNA | AK014477.1 | - | 13 | 1464 | |
AK014795.1 | - | 8 | 915 | ||
AK078875.1 | - | 13 | 1478 | ||
AK135905.1 | - | 9 | 1008 | ||
AK147085.1 | - | 13 | 1495 | ||
AK162115.1 | - | 2 | 130 | ||
BC032087.1 | - | 13 | 1460 | ||
BC053395.1 | - | 13 | 1513 | 30054233 | |
DQ054831.1 | - | 13 | 1463 |