Gene Transcripts
- Gene Symbol
- Dnajc19
- Entrez Gene ID
- 67713
- Gene Name
- DnaJ (Hsp40) homolog, subfamily C, member 19
- Gene Aliases
- 1810055D05Rik, AA959924, Tim14
- Chromosome Location
- Chr. 3: 34057280 - 34081354 on Build GRCm38
- UniGene ID
- Mm.274266
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001026211.2 |
NP_001021382.1
|
2, 3 |
219 |
|
NM_001286972.1 |
NP_001273901.1
|
2, 3 |
219 |
|
NM_026332.4 |
NP_080608.3
|
2, 3 |
219 |
|
GenBank mRNA |
AK007866.1 |
- |
2, 3 |
187 |
|
AK008267.1 |
- |
2, 3 |
163 |
|
AK010204.1 |
- |
2, 3 |
169 |
|
AK012051.1 |
- |
2, 3 |
171 |
|
AK076602.1 |
- |
|
178 |
|
BC020327.1 |
- |
2, 3 |
152 |
|
BC024335.1 |
- |
2, 3 |
127 |
4974988 |
BY149679.1 |
- |
2, 3 |
192 |
|
CA469052.1 |
- |
2, 3 |
256 |
|
EL962106.1 |
- |
2, 3 |
165 |
|
View Full Product Details