Gene Transcripts

Gene Symbol
Tti1
Entrez Gene ID
75425
Gene Name
TELO2 interacting protein 1
Gene Aliases
2610036D13Rik, AI449463
Chromosome Location
Chr. 2: 157981803 - 158028477 on Build GRCm38
UniGene ID
Mm.29068
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029282.1 NP_083558.1 4, 5 2819
GenBank mRNA AF332067.1 - 4, 5 2802
AF332068.1 - 4, 5 2802
AK128940.1 - 5, 6 2869
BC053067.1 - 4, 5 2819 5698252

View Full Product Details