Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081287.2 | NP_001074756.2 | 11 | 1176 | |
NM_001161620.1 | NP_001155092.1 | 11 | 1176 | ||
GenBank mRNA | AK012883.1 | - | 11 | 1151 | |
AK033081.1 | - | 11 | 1136 | ||
AK036956.1 | - | 11 | 1147 | ||
AK078849.1 | - | 11 | 1175 | ||
BC049662.1 | - | 11 | 1130 | ||
BC117550.1 | - | 10 | 1023 | 40105182 | |
BC118058.1 | - | 10 | 1020 | 40105180 |