Gene Transcripts

Gene Symbol
Samd7
Entrez Gene ID
75953
Gene Name
sterile alpha motif domain containing 7
Gene Aliases
4930597A01Rik
Chromosome Location
Chr. 3: 30746293 - 30767174 on Build GRCm38
UniGene ID
Mm.308084
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029489.3 NP_083765.2 5 936
GenBank mRNA AK016401.1 - 1 431
AK044227.1 - 4 812

View Full Product Details