Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153196.1 | NP_694876.1 | 6 | 582 | |
GenBank mRNA | AK017229.1 | - | 6 | 600 | |
AK138939.1 | - | 6 | 600 | ||
BC023339.1 | - | 6 | 582 | 5253827 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF453934.1 | - | 441 | Chr. - - [-] |