Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001191008.1 | NP_001177937.1 | 2 | 853 | |
NM_011425.3 | NP_035555.1 | 3 | 1005 | ||
GenBank mRNA | AK133609.1 | - | 3 | 971 | |
AK133767.1 | - | 2 | 781 | ||
BC145943.1 | - | 3 | 1005 | 40131246 | |
BC150805.1 | - | 2 | 837 | 9087716 |