Gene Transcripts

Gene Symbol
Spink1
Entrez Gene ID
20730
Gene Name
serine peptidase inhibitor, Kazal type 1
Gene Aliases
Spink3, p12
Chromosome Location
Chr. 18: 43728069 - 43737237 on Build GRCm38
UniGene ID
Mm.272
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009258.5 NP_033284.1 1, 2 133
GenBank mRNA AK007841.1 - 1, 2 94
AK007985.1 - 1, 2 92
BC086887.1 - 1, 2 92 6775994
CA481615.1 - 1, 2 170
X06342.1 - 1, 2 90

View Full Product Details