Gene Transcripts

Gene Symbol
Serpine3
Entrez Gene ID
319433
Gene Name
serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 3
Gene Aliases
E130113E03Rik
Chromosome Location
Chr. 14: 62663667 - 62692243 on Build GRCm38
UniGene ID
Mm.483827
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199945.1 NP_001186874.1 4 702
GenBank mRNA AK053602.1 - 4 704

View Full Product Details