Gene Transcripts

Gene Symbol
Utp11l
Entrez Gene ID
67205
Gene Name
UTP11-like, U3 small nucleolar ribonucleoprotein, (yeast)
Gene Aliases
2700082D03Rik, CGI-94, Cgi94
Chromosome Location
Chr. 4: 124678232 - 124693612 on Build GRCm38
UniGene ID
Mm.259121
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026031.3 NP_080307.1 6 537
GenBank mRNA AK007361.1 - 4 308
AK008801.1 - 2 89
AK012550.1 - 6 538
AK145266.1 - 6 570
AK147113.1 - 6 568
BC043139.1 - 5 1245
BC061074.1 - 6 566 6706808

View Full Product Details