Gene Transcripts

Gene Symbol
Dydc2
Entrez Gene ID
71200
Gene Name
DPY30 domain containing 2
Gene Aliases
4933428D01Rik
Chromosome Location
Chr. 14: 41048957 - 41074049 on Build GRCm38
UniGene ID
Mm.443973
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027717.1 NP_081993.1 4 469
GenBank mRNA AK016963.1 - 4 604
BC113750.1 - 4 469 40049377

View Full Product Details