Gene Transcripts

Gene Symbol
Zdhhc8
Entrez Gene ID
303796
Gene Name
zinc finger, DHHC-type containing 8
Gene Aliases
Chromosome Location
Chr. 11: 86903122 - 86917261 on Build Rnor_6.0
UniGene ID
Rn.17261
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039021.1 NP_001034110.1 4 563
XM_006248610.2 XP_006248672.1 4 562
GenBank mRNA AY871204.1 - 4 563

View Full Product Details