Gene Transcripts

Gene Symbol
Xpo5
Entrez Gene ID
363194
Gene Name
exportin 5
Gene Aliases
Exp5, RanBp21
Chromosome Location
Chr. 9: 17125201 - 17163170 on Build Rnor_6.0
UniGene ID
Rn.19170
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108789.3 NP_001102259.2 21 2480
GenBank mRNA DN935088.1 - 4 444

View Full Product Details