Gene Transcripts

Gene Symbol
Vom1r97
Entrez Gene ID
286956
Gene Name
vomeronasal 1 receptor 97
Gene Aliases
V1rb12, V1rb9, Vmn1r41, Vn5, Vnr5
Chromosome Location
Chr. 4: 121899292 - 121900224 on Build Rnor_6.0
UniGene ID
Rn.10143
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173297.2 NP_775419.2 1 649
GenBank mRNA AY510281.1 - 1 649
U36897.1 - 1 555

View Full Product Details