Gene Transcripts

Gene Symbol
Vom1r87
Entrez Gene ID
494249
Gene Name
vomeronasal 1 receptor 87
Gene Aliases
V1rc17
Chromosome Location
Chr. 4: 88328061 - 88328969 on Build Rnor_6.0
UniGene ID
Rn.129653
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009529.1 NP_001009529.1 1 118
GenBank mRNA AY510311.1 - 1 118

View Full Product Details