Gene Transcripts
- Gene Symbol
- Olfr75-ps1
- Entrez Gene ID
- 258186
- Gene Name
- olfactory receptor 75, pseudogene 1
- Gene Aliases
- MOR135-20, Olfr75, V1
- Chromosome Location
- Chr. 11: 73405450 - 73409858 on Build GRCm38
- UniGene ID
- Mm.173657
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002859.2 |
|
2 |
764 |
|
NR_033528.1 |
|
3 |
983 |
|
GenBank mRNA |
Y15524.1 |
- |
1 |
462 |
|
View Full Product Details