Gene Transcripts

Gene Symbol
Tbrg3
Entrez Gene ID
21378
Gene Name
transforming growth factor beta regulated gene 3
Gene Aliases
TB-11
Chromosome Location
Chr. 15: 82890483 - 82898461 on Build GRCm38
UniGene ID
Mm.417272
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027799.1 1 285
GenBank mRNA AK042509.1 - 1 286
AK090121.1 - 1611
BC095996.1 - 1 192

View Full Product Details