Gene Transcripts

Gene Symbol
Mir22hg
Entrez Gene ID
100042498
Gene Name
Mir22 host gene (non-protein coding)
Gene Aliases
2210403K04Rik, mir-22
Chromosome Location
Chr. 11: 75461539 - 75466690 on Build GRCm38
UniGene ID
Mm.482242
Species
Mus musculus
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_030711.1
3 1499
GenBank mRNA AK008813.1 - LIT3441
2210403K04Rik
3 1391
AK018628.1 -
3 1409

View Full Product Details