Gene Transcripts
- Gene Symbol
- SNORA61
- Entrez Gene ID
- 677838
- Gene Name
- small nucleolar RNA, H/ACA box 61
- Gene Aliases
- ACA61
- Chromosome Location
- Chr. 1: 28579764 - 28579893 on Build GRCh38
- UniGene ID
- Hs.632377
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002987.1 |
|
1 |
77 |
|
GenBank mRNA |
AJ609451.1 |
- |
1 |
77 |
|
- Gene Symbol
- SNHG12
- Entrez Gene ID
- 85028
- Gene Name
- small nucleolar RNA host gene 12
- Gene Aliases
- ASLNC04080, C1orf79, LINC00100, NCRNA00100
- Chromosome Location
- Chr. 1: 28578538 - 28581854 on Build GRCh38
- UniGene ID
- Hs.632377
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK092096.1 |
- |
2 |
1481 |
|
BC045792.1 |
- |
4 |
671 |
|
BC064589.1 |
- |
3 |
1202 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CA495298.1 |
- |
4 |
642 |
|
Chr. 1 - 28579641 - 28581859 [-] |
GenBankmRNA |
DR002536.1 |
- |
4 |
647 |
|
Chr. 1 - 28579688 - 28581865 [-] |
View Full Product Details