Gene Transcripts

Gene Symbol
OR7E47P
Entrez Gene ID
26628
Gene Name
olfactory receptor family 7 subfamily E member 47 pseudogene
Gene Aliases
OR7E141
Chromosome Location
Chr. 12: 52079696 - 52108319 on Build GRCh38
UniGene ID
Hs.524431
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_120437.1 3 815
NR_120439.1 2 338
NR_120440.1 2 315
GenBank mRNA BC042060.1 - 3 815

View Full Product Details