Gene Transcripts
- Gene Symbol
- TRAM2
- Entrez Gene ID
- 9697
- Gene Name
- translocation associated membrane protein 2
- Gene Aliases
- Chromosome Location
- Chr. 6: 52497402 - 52577064 on Build GRCh38
- UniGene ID
- Hs.520182
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY927606.1 |
- |
LIT3596 AY927606 |
2 |
198 |
|
View Full Product Details