Gene Transcripts
- Gene Symbol
- SPINK13
- Entrez Gene ID
- 153218
- Gene Name
- serine peptidase inhibitor, Kazal type 13 (putative)
- Gene Aliases
- HBVDNAPTP1, HESPINTOR, LiESP6, SPINK5L3
- Chromosome Location
- Chr. 5: 148268860 - 148286210 on Build GRCh38
- UniGene ID
- Hs.483771
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK001520.1 |
- |
1 |
89 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AU129688.1 |
- |
1 |
89 |
|
Chr. 5 - 148268180 - 148282204 [+] |
View Full Product Details