Gene Transcripts
- Gene Symbol
- PRKXP1
- Entrez Gene ID
- 441733
- Gene Name
- protein kinase, X-linked, pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 15: 100547752 - 100559283 on Build GRCh38
- UniGene ID
- Hs.12250
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_073405.1 |
|
|
10802 |
|
GenBank mRNA |
AK095793.1 |
- |
1 |
1677 |
|
Y10481.1 |
- |
1 |
463 |
|
View Full Product Details